Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-3-amino-4-fluorobenzoate 95%

Supplier: AMBEED, INC
Warning

A212810-1G A212810-5G A212810-1G A212810-25G A212810-5G
103818-240EA 7 USD InStock
103818-240 103848-936 77087-408 77087-410 77087-412
Ethyl-3-amino-4-fluorobenzoate 95%
Ethyl-3-amino-4-fluorobenzoate
Precaución: for Research Use Only


Formula: C₉H₁₀FNO₂
MW: 183.1796 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03840487
CAS Number: 455-75-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty