Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 3-bromo-1-methyl-1H-pyrazole-4-carboxylate 95%

Supplier: AMBEED, INC
Warning

A211611-1G A211611-5G A211611-250MG A211611-1G
103818-124EA 245 USD InStock
103818-124 103848-818 77237-398 77303-202
Ethyl 3-bromo-1-methyl-1H-pyrazole-4-carboxylate 95%
Ethyl 3-bromo-1-methyl-1H-pyrazole-4-carboxylate
Precaución: for Research Use Only


Formula: C₇H₉BrN₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD22420858
CAS Number: 139308-52-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty