Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 3-fluoro-1H-pyrrole-2-carboxylate 95%

Supplier: AMBEED, INC
Warning

A136441-1G A136441-250MG A136441-5G
103802-300EA 39 USD InStock
103802-300 76811-548 77298-850
Ethyl 3-fluoro-1H-pyrrole-2-carboxylate 95%
Ethyl 3-fluoro-1H-pyrrole-2-carboxylate
Precaución: for Research Use Only


Formula: C₇H₈FNO₂
MW: 157.142 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD11042605
CAS Number: 168102-05-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty