Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 3-iodo-1H-indazole-5-carboxylate 97%

Supplier: AMBEED, INC
Warning

A405856-1G A405856-100MG A405856-250MG
103827-124EA 39 USD InStock
103827-124 103838-164 76841-656 77168-346 77168-348
Ethyl 3-iodo-1H-indazole-5-carboxylate 97%
Ethyl 3-iodo-1H-indazole-5-carboxylate
Precaución: for Research Use Only


Formula: C₁₀H₉IN₂O₂
Boiling Pt: 427.6±25.0 °C (760 mm Hg)
Flash Pt: 212.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD23704369
CAS Number: 1279863-38-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty