Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 3-methoxy-1H-pyrazole-4-carboxylate 96%

Supplier: AMBEED, INC
Warning

A417704-5G A417704-1G A417704-100MG A417704-250MG
103827-338EA 101 USD InStock
103827-338 103838-384 76841-974 77168-486 77168-488
Ethyl 3-methoxy-1H-pyrazole-4-carboxylate 96%
Ethyl 3-methoxy-1H-pyrazole-4-carboxylate
Precaución: for Research Use Only


Formula: C₇H₁₀N₂O₃
MW: 170.17 g/mol
Melting Pt: 68…71 °C
Storage Temperature: Ambient
MDL Number: MFCD13689107
CAS Number: 478968-48-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty