Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-3-oxocyclopentane-1-carboxylate 98%

Supplier: AMBEED, INC
Warning

A118711-1G A118711-10G A118711-1G A118711-5G A118711-25G
103799-098EA 14 USD InStock
103799-098 76797-242 77311-768 77311-770 77878-886
Ethyl-3-oxocyclopentane-1-carboxylate 98%
Ethyl-3-oxocyclopentane-1-carboxylate
Precaución: for Research Use Only


Formula: C₈H₁₂O₃
Boiling Pt: 228.2 °C (760 mm Hg)
Flash Pt: 93.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01320352
CAS Number: 5400-79-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty