Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 4-(2-nitrophenyl)-3-oxobutanoate 98%

Supplier: AMBEED, INC
Warning

A155253-1G A155253-5G A155253-100MG A155253-250MG A155253-1G
103804-736EA 125 USD InStock
103804-736 103842-930 76798-778 76798-780 77300-106
Ethyl 4-(2-nitrophenyl)-3-oxobutanoate 98%
Ethyl 4-(2-nitrophenyl)-3-oxobutanoate
Precaución: for Research Use Only


Formula: C₁₂H₁₃NO₅
Storage Temperature: Ambient
MDL Number: MFCD01926658
CAS Number: 66073-33-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty