Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-4,4-difluoroacetoacetate 97% (by GC)

Supplier: AMBEED, INC
Warning

A129869-100G A129869-500G
76797-832EA 39 USD InStock
76797-832 76797-834 76797-836
Ethyl-4,4-difluoroacetoacetate 97% (by GC)
Ethyl-4,4-difluoroacetoacetate
Precaución: for Research Use Only


Formula: C₆H₈F₂O₃
MW: 166.12 g/mol
Boiling Pt: 170.1 °C (760 mm Hg)
Flash Pt: 68.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00039277
CAS Number: 352-24-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty