Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 4,5-dimethyl-1H-pyrazole-3-carboxylate 95%

Supplier: AMBEED, INC
Warning

A229592-1G A229592-5G A229592-250MG A229592-1G
103819-484EA 266 USD InStock
103819-484 103850-386 76987-292 77303-852
Ethyl 4,5-dimethyl-1H-pyrazole-3-carboxylate 95%
Ethyl 4,5-dimethyl-1H-pyrazole-3-carboxylate
Precaución: for Research Use Only


Formula: C₈H₁₂N₂O₂
MW: 168.19 g/mol
Storage Temperature: Ambient
MDL Number: MFCD22572310
CAS Number: 15803-27-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty