Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-4-acetylbenzoate 95%

Supplier: AMBEED, INC
Warning

A171850-1G A171850-5G A171850-100MG A171850-10G A171850-250MG A171850-1G A171850-25G A171850-5G
103806-672EA 8 USD InStock
103806-672 103844-952 76799-440 76799-442 76799-444 77082-124 77082-126 77082-128
Ethyl-4-acetylbenzoate 95%
Ethyl-4-acetylbenzoate
Precaución: for Research Use Only


Formula: C₁₁H₁₂O₃
Boiling Pt: 310 °C (760 mm Hg)
Melting Pt: 55…57 °C
Flash Pt: 135.8 °C
Storage Temperature: Ambient
MDL Number: MFCD00013241
CAS Number: 38430-55-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty