Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 4-hydrazinylbenzoate 95%

Supplier: AMBEED, INC
Warning

A267394-1G A267394-5G A267394-250MG A267394-1G A267394-5G
103812-850EA 123 USD InStock
103812-850 103852-748 76787-118 77092-482 77092-484
Ethyl 4-hydrazinylbenzoate 95%
Ethyl 4-hydrazinylbenzoate
Precaución: for Research Use Only


Formula: C₉H₁₂N₂O₂
MW: 180.2 g/mol
Flash Pt: 153.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00278571
CAS Number: 14685-90-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty