Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl vanillate 98%

Supplier: AMBEED, INC
Warning

A202337-5G A202337-100MG A202337-10G A202337-25G A202337-100G
103848-012EA 6 USD InStock
103848-012 76691-250 76691-252 77086-120 77626-032
Ethyl vanillate 98%
Ethyl vanillate
Precaución: for Research Use Only


Formula: C₁₀H₁₂O₄
MW: 196.20 g/mol
Boiling Pt: 292 °C (760 mm Hg)
Melting Pt: 39…41 °C
Flash Pt: 122.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00017269
CAS Number: 617-05-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty