Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 4-iodo-1H-pyrazole-5-carboxylate 98%

Supplier: AMBEED, INC
Warning

A147771-1G A147771-5G A147771-10G A147771-250MG A147771-1G A147771-25G A147771-5G
103803-832EA 11 USD InStock
103803-832 103841-940 76823-714 76823-716 77317-340 77317-342 77317-344
Ethyl 4-iodo-1H-pyrazole-5-carboxylate 98%
Ethyl 4-iodo-1H-pyrazole-5-carboxylate
Precaución: for Research Use Only


Formula: C₆H₇IN₂O₂
Boiling Pt: 345.6±22.0 °C
Flash Pt: 162.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00466329
CAS Number: 179692-08-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty