Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-5-bromo-1H-1,2,4-triazole-3-carboxylate 98%

Supplier: AMBEED, INC
Warning

A115332-1G A115332-250MG A115332-1G A115332-5G A115332-25G A115332-10G
103798-548EA 17 USD InStock
103798-548 103824-402 77108-238 77311-214 77311-216 77685-022 77685-021
Ethyl-5-bromo-1H-1,2,4-triazole-3-carboxylate 98%
Ethyl-5-bromo-1H-1,2,4-triazole-3-carboxylate
Precaución: for Research Use Only


Formula: C₅H₆BrN₃O₂
MW: 220.0240 g/mol
Flash Pt: 167.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD11040185
CAS Number: 774608-89-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty