Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-5-bromovalerate 98%

Supplier: AMBEED, INC
Warning

A104682-5G A104682-100G A104682-10G A104682-25G A104682-500G
103822-480EA 8 USD InStock
103822-480 76994-770 76994-772 76994-774 76994-776
Ethyl-5-bromovalerate 98%
Ethyl-5-bromovalerate
Precaución: for Research Use Only


Formula: BrH₂C(CH₂)₃COOCH₂CH₃
MW: 209.08 g/mol
Boiling Pt: 104
Flash Pt: 104 °C
Storage Temperature: Ambient
MDL Number: MFCD00000266
CAS Number: 14660-52-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty