Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-5-methyl-1H-imidazole-4-carboxylate 98%

Supplier: AMBEED, INC
Warning

A211377-1G A211377-5G A211377-10G A211377-500G A211377-1G A211377-25G A211377-5G
103818-102EA 10 USD InStock
103818-102 103848-796 76975-558 76975-560 77087-200 77303-190 77303-192
Ethyl-5-methyl-1H-imidazole-4-carboxylate 98%
Ethyl-5-methyl-1H-imidazole-4-carboxylate
Precaución: for Research Use Only


Formula: C₇H₁₀N₂O₂
Boiling Pt: 323.6 °C (760 mm Hg)
Melting Pt: 204…206 °C
Flash Pt: 149.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00005199
CAS Number: 51605-32-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty