Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 6-(trifluoromethyl)-1H-indole-2-carboxylate 96%

Supplier: AMBEED, INC
Warning

A111063-1G A111063-5G A111063-250MG A111063-1G A111063-5G A111063-100MG
103797-786EA 289 USD InStock
103797-786 103823-596 76796-850 77310-404 77310-406 77878-841
Ethyl 6-(trifluoromethyl)-1H-indole-2-carboxylate 96%
Ethyl 6-(trifluoromethyl)-1H-indole-2-carboxylate
Precaución: for Research Use Only


Formula: C₁₂H₁₀F₃NO₂
MW: 257.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09833401
CAS Number: 327-21-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty