Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-6,8-dichlorooctanoate 95%

Supplier: AMBEED, INC
Danger

A586355-1G A586355-25G A586355-5G A586355-100G A586355-10G A586355-1G A586355-1000G
103828-888EA 5 USD InStock
103828-888 103840-036 76844-174 76844-176 76980-420 76980-422 77307-878 77882-393
Ethyl-6,8-dichlorooctanoate 95%
Ethyl-6,8-dichlorooctanoate
Precaución: For research use only


Formula: C₁₀H₁₈Cl₂O₂
Boiling Pt: 288.5 °C (760 mm Hg)
Flash Pt: 110 °C
Storage Temperature: Ambient
MDL Number: MFCD01711127
CAS Number: 1070-64-0
UN: 3082
ADR: 9,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty