Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl 6-chloropyrimidine-4-carboxylate 97%

Supplier: AMBEED, INC
Warning

A211783-1G A211783-5G A211783-10G A211783-250MG A211783-1G A211783-5G A211783-100G
103818-136EA 16 USD InStock
103818-136 103848-832 76975-570 76975-572 77303-210 77303-212 77881-510
Ethyl 6-chloropyrimidine-4-carboxylate 97%
Ethyl 6-chloropyrimidine-4-carboxylate
Precaución: for Research Use Only


Formula: C₇H₇ClN₂O₂
Melting Pt: 65…69 °C
Storage Temperature: Refrigerator
MDL Number: MFCD12756023
CAS Number: 51940-63-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty