Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-7-aminofuro[2,3-b]pyrazine-6-carboxylate 95%

Supplier: AMBEED, INC
Warning

A310743-1G A310743-250MG A310743-1G A310743-5G A310743-100MG
103814-634EA 66 USD InStock
103814-634 103835-818 76787-906 76838-590 76838-592 77872-429
Ethyl-7-aminofuro[2,3-b]pyrazine-6-carboxylate 95%
Ethyl-7-aminofuro[2,3-b]pyrazine-6-carboxylate
Precaución: for Research Use Only


Formula: C₉H₉N₃O₃
Boiling Pt: 345.6 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD09909670
CAS Number: 187732-95-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty