Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl isoquinoline-7-carboxylate 97%

Supplier: AMBEED, INC
Warning

A100457-100MG A100457-1G A100457-250MG
103795-450EA 257 USD InStock
103795-450 76821-202 76821-204 76821-206
Ethyl isoquinoline-7-carboxylate 97%
Ethyl isoquinoline-7-carboxylate
Precaución: for Research Use Only


Formula: C₁₂H₁₁NO₂
MW: 201.22 g/mol
Boiling Pt: 341.8±15.0 °C (760 mm Hg)
Flash Pt: 160.5 °C
Storage Temperature: Ambient
MDL Number: MFCD18416670
CAS Number: 407623-83-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty