Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl morpholine-2-carboxylate 96%

Supplier: AMBEED, INC
Warning

A145744-1G A145744-5G A145744-1G A145744-250MG
103803-578EA 37 USD InStock
103803-578 103841-660 77316-968 77871-714
Ethyl morpholine-2-carboxylate 96%
Ethyl morpholine-2-carboxylate
Precaución: for Research Use Only


Formula: C₇H₁₃NO₃
MW: 159.19 g/mol
Boiling Pt: 221.4 °C (760 mm Hg)
Flash Pt: 87.702 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08751336
CAS Number: 135782-25-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty