Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-4-piperidinecarboxylate 97%

Supplier: AMBEED, INC
Warning

A132547-100G A132547-500G A132547-25G A132547-5G A132547-10G
77190-112EA 55 USD InStock
77190-112 77190-114 77314-328 77314-330 77592-526
Ethyl-4-piperidinecarboxylate 97%
Ethyl-4-piperidinecarboxylate
Precaución: for Research Use Only


Formula: C₈H₁₅NO₂
MW: 157.21 g/mol
Boiling Pt: 204 °C (760 mm Hg)
Flash Pt: 80 °C
Storage Temperature: Ambient
MDL Number: MFCD00006003
CAS Number: 1126-09-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty