Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl pyrimidine-2-carboxylate 98%

Supplier: AMBEED, INC
Warning

A238517-1G A238517-5G A238517-10G A238517-250MG A238517-1G
103811-234EA 9 USD InStock
103811-234 103851-016 76826-672 76826-674 77304-110
Ethyl pyrimidine-2-carboxylate 98%
Ethyl pyrimidine-2-carboxylate
Precaución: for Research Use Only


Formula: C₇H₈N₂O₂
Storage Temperature: Ambient
MDL Number: MFCD12197317
CAS Number: 42839-08-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty