Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fenofibric acid 98%

Supplier: AMBEED, INC
Warning

A399267-5G A399267-25G A399267-100G A399267-10G A399267-500G A399267-5G A399267-1000G
103838-064EA 5 USD InStock
103838-064 76841-500 77252-104 77252-106 77252-108 77252-110 77307-060 77746-802
Fenofibric acid 98%
Fenofibric acid
Precaución: for Research Use Only


Formula: C₁₇H₁₅ClO₄
MW: 318.75 g/mol
Boiling Pt: 486.5 °C (760 mm Hg)
Melting Pt: 177…179 °C
Flash Pt: 248 °C
Storage Temperature: Ambient
MDL Number: MFCD00792461
CAS Number: 42017-89-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty