Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Flibanserin Hydrochloride 95%

Supplier: AMBEED, INC
Warning

A109623-1G A109623-5G A109623-10G A109623-1G A109623-5G
103797-534EA 78 USD InStock
103797-534 103823-334 77093-910 77310-146 77310-148
Flibanserin Hydrochloride 95%
Flibanserin Hydrochloride
Precaución: for Research Use Only


Formula: C₂₀H₂₂ClF₃N₄O
Melting Pt: >225 °C
Storage Temperature: Ambient
MDL Number: MFCD29058559
CAS Number: 147359-76-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty