Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Flumequine 98%

Supplier: AMBEED, INC
Warning

A127261-1G A127261-100MG A127261-25G A127261-50MG A127261-100G
103800-942EA 11 USD InStock
103800-942 76797-710 77666-818 77878-929 77878-928
Flumequine 98%
Flumequine
Precaución: for Research Use Only


Formula: C₁₄H₁₂FNO₃
Boiling Pt: 439.7 °C (760 mm Hg)
Melting Pt: 253…255 °C
Flash Pt: 219.7 °C
Storage Temperature: Freezer
MDL Number: MFCD00079298
CAS Number: 42835-25-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty