Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-[[(9H-Fluoren-9-ylmethoxy)carbonyl]aminomethyl]benzoic acid 97%

Supplier: AMBEED, INC
Warning

A262319-1G A262319-1G A262319-25G A262319-5G A262319-100G A262319-10G
103812-558EA 5 USD InStock
103812-558 103852-454 77092-068 77092-070 77092-072 77289-234 77289-236
4-[[(9H-Fluoren-9-ylmethoxy)carbonyl]aminomethyl]benzoic acid 97%
4-[[(9H-Fluoren-9-ylmethoxy)carbonyl]aminomethyl]benzoic acid
Precaución: for Research Use Only


Formula: C₂₃H₁₉NO₄
Boiling Pt: 625.4 °C (760 mm Hg)
Melting Pt: 224…228 °C
Flash Pt: 332 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01074693
CAS Number: 164470-64-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty