Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fmoc-α-methylalanine 98%

Supplier: AMBEED, INC
Warning

A142078-100G A142078-10G A142078-500G A142078-25G
103808-624EA 629 USD InStock
103808-624 77109-850 77109-852 77109-854 77316-308
Fmoc-α-methylalanine 98%
Fmoc-alpha-methylalanine
Precaución: for Research Use Only


Formula: C₁₉H₁₉NO₄
MW: 325.364 g/mol
Boiling Pt: 544.3 °C (760 mm Hg)
Melting Pt: 182…188 °C
Flash Pt: 283 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00151913
CAS Number: 94744-50-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty