Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fmoc-Arg-OH 97%

Supplier: AMBEED, INC
Warning

A273522-1G A273522-5G A273522-100G A273522-1G A273522-25G A273522-10G
103813-146EA 6 USD InStock
103813-146 103853-066 76723-092 77305-136 77305-138 77668-194
Fmoc-Arg-OH 97%
Fmoc-Arg-OH
Precaución: for Research Use Only


Formula: C₂₁H₂₄N₄O₄
Melting Pt: 145…150 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00051770
CAS Number: 91000-69-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty