Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fmoc-Asp-OAll 95%

Supplier: AMBEED, INC
Warning

A176715-1G A176715-5G A176715-10G A176715-250MG A176715-1G A176715-25G A176715-5G
103807-170EA 20 USD InStock
103807-170 103845-496 76998-390 76998-392 77082-850 77082-852 77082-854
Fmoc-Asp-OAll 95%
Fmoc-Asp-OAll
Precaución: for Research Use Only


Formula: C₂₂H₂₁NO₆
MW: 395.411 g/mol
Boiling Pt: 634.8 °C (760 mm Hg)
Melting Pt: 117…139 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00467715
CAS Number: 144120-53-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty