Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fmoc-Dab(Boc)-OH 97%

Supplier: AMBEED, INC
Warning

A239334-1G A239334-5G A239334-1G A239334-5G A239334-10G A239334-250MG
103811-278EA 7 USD InStock
103811-278 103851-076 77090-218 77090-220 77153-048 77153-050
Fmoc-Dab(Boc)-OH 97%
Fmoc-Dab(Boc)-OH
Precaución: for Research Use Only


Formula: C₂₄H₂₈N₂O₆
MW: 440.496 g/mol
Boiling Pt: 670.9 °C (760 mm Hg)
Melting Pt: 111…113 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00151941
CAS Number: 125238-99-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty