Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fmoc-D-Asp-OMe 97%

Supplier: AMBEED, INC
Warning

A195314-1G A195314-5G A195314-1G A195314-100G A195314-10G A195314-250MG A195314-5G
103816-742EA 6 USD InStock
103816-742 103847-356 77085-304 77139-224 77139-226 77139-228 77302-456
Fmoc-D-Asp-OMe 97%
Fmoc-D-Asp-OMe
Precaución: for Research Use Only


Formula: C₂₀H₁₉NO₆
MW: 369.36 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD09842078
CAS Number: 368443-82-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty