Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Nε-(tert-butoxycarbonyl)-Nɑ-[(9H-fluoren-9-ylmethoxy)carbonyl]-D-lysine 98%

Supplier: AMBEED, INC
Warning

A141375-1G A141375-100G A141375-10G A141375-1G A141375-25G A141375-5G
103802-984EA 9 USD InStock
103802-984 76871-214 76871-216 77316-138 77316-140 77316-142
Nε-(tert-butoxycarbonyl)-Nɑ-[(9H-fluoren-9-ylmethoxy)carbonyl]-D-lysine 98%
Nepsilon-(tert-butoxycarbonyl)-Nalpha-[(9H-fluoren-9-ylmethoxy)carbonyl]-D-lysine
Precaución: for Research Use Only


Formula: C₂₆H₃₂N₂O₆
MW: 468.55 g/mol
Melting Pt: 128…131 °C
Flash Pt: 368.5 °C
Storage Temperature: Ambient
MDL Number: MFCD00065660
CAS Number: 92122-45-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty