Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fmoc-D-propargylglycine 97%

Supplier: AMBEED, INC
Warning

A156735-1G A156735-5G A156735-10G A156735-250MG A156735-25G A156735-5G
103804-918EA 32 USD InStock
103804-918 103843-118 76997-560 76997-562 77300-208 77300-210
Fmoc-D-propargylglycine 97%
Fmoc-D-propargylglycine


Formula: C₂₀H₁₇NO₄
Boiling Pt: 417 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD01311781
CAS Number: 220497-98-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty