Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fmoc-D-Thr(tBu)-OH 97%

Supplier: AMBEED, INC
Warning

A183224-1G A183224-5G A183224-10G A183224-250MG A183224-1G A183224-25G A183224-5G
103815-620EA 7 USD InStock
103815-620 103846-182 76784-658 76784-660 77301-824 77301-826 77301-828
Fmoc-D-Thr(tBu)-OH 97%
Fmoc-D-Thr(tBu)-OH
Precaución: for Research Use Only


Formula: C₂₃H₂₇NO₅
MW: 397.471 g/mol
Boiling Pt: 581.7 °C (760 mm Hg)
Melting Pt: 130 °C
Flash Pt: 305.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00080292
CAS Number: 138797-71-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty