Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fmoc-Glu-OAll 97%

Supplier: AMBEED, INC
Warning

A178277-1G A178277-5G A178277-100G A178277-25G A178277-1G A178277-5G
103807-310EA 15 USD InStock
103807-310 103845-650 76998-480 77083-058 77301-548 77301-550
Fmoc-Glu-OAll 97%
Fmoc-Glu-OAll
Precaución: For research use only.


Formula: C₂₃H₂₃NO₆
MW: 409.438 g/mol
Boiling Pt: 652.3 °C (760 mm Hg)
Melting Pt: 118…122 °C
Flash Pt: 348.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00467718
CAS Number: 144120-54-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty