Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fmoc-Glutamol(OtBu) 95%

Supplier: AMBEED, INC
Warning

A241105-1G A241105-5G A241105-10G A241105-250MG A241105-1G A241105-5G
103811-394EA 36 USD InStock
103811-394 103851-200 76801-344 76801-346 77090-364 77090-366
Fmoc-Glutamol(OtBu) 95%
Fmoc-Glutamol(OtBu)
Precaución: for Research Use Only


Formula: C₂₄H₂₉NO₅
Boiling Pt: 590.1±50.0 °C (760 mm Hg)
Melting Pt: 57…59 °C
Flash Pt: 310.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00235926
CAS Number: 153815-59-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty