Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fmoc-Gly-Osu 95%

Supplier: AMBEED, INC
Warning

A133418-1G A133418-100G A133418-10G A133418-250MG A133418-1G A133418-25G A133418-5G
103801-876EA 6 USD InStock
103801-876 76996-512 76996-514 76996-516 77298-632 77314-526 77314-528
Fmoc-Gly-Osu 95%
Fmoc-Gly-Osu
Precaución: for Research Use Only


Formula: C₂₁H₁₈N₂O₆
MW: 394.383 g/mol
Melting Pt: 173…175 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00065651
CAS Number: 113484-74-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty