Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fmoc-Lys(Ddiv)-OH 96%

Supplier: AMBEED, INC
Warning

A435571-1G A435571-5G A435571-100G A435571-250MG
103827-598EA 95 USD InStock
103827-598 103838-666 76842-390 76842-392 77102-366 77102-368
Fmoc-Lys(Ddiv)-OH 96%
Fmoc-Lys(Ddiv)-OH
Precaución: for Research Use Only


Formula: C₃₄H₄₂N₂O₆
MW: 574.718 g/mol
Boiling Pt: 765.6 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD01631658
CAS Number: 204777-78-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty