Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fmoc-Lys-OH 97%

Supplier: AMBEED, INC
Warning

A186089-1G A186089-5G A186089-100G A186089-10G A186089-5G A186089-1G A186089-25G
103815-910EA 6 USD InStock
103815-910 103846-484 76998-842 76998-844 77084-132 77302-018 77302-020
Fmoc-Lys-OH 97%
Fmoc-Lys-OH
Precaución: for Research Use Only


Formula: C₂₁H₂₄N₂O₄
MW: 368.44 g/mol
Boiling Pt: 607.6 °C (760 mm Hg)
Flash Pt: 321.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00038539
CAS Number: 105047-45-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty