Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fmoc-L-β-homoleucine 98%

Supplier: AMBEED, INC
Warning

A500874-1G A500874-5G A500874-100MG A500874-250MG
103828-290EA 40 USD InStock
103828-290 103839-378 77004-686 77004-688
Fmoc-L-β-homoleucine 98%
Fmoc-L-beta-homoleucine
Precaución: for Research Use Only


Formula: C₂₂H₂₅NO₄
Boiling Pt: 572.1 °C (760 mm Hg)
Flash Pt: 299.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01863059
CAS Number: 193887-44-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty