Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fmoc-N-Me-Asp(OtBu)-OH 98%

Supplier: AMBEED, INC
Warning

A208912-1G A208912-5G A208912-250MG A208912-5G A208912-1G A208912-100MG A208912-25G A208912-10G
103817-896EA 94 USD InStock
103817-896 103848-584 77030-448 77086-916 77303-078 77667-756 77667-758 77872-058
Fmoc-N-Me-Asp(OtBu)-OH 98%
Fmoc-N-Me-Asp(OtBu)-OH
Precaución: for Research Use Only


Formula: C₂₄H₂₇NO₆
MW: 425.481 g/mol
Boiling Pt: 598.7 °C (760 mm Hg)
Melting Pt: 135…140 °C
Flash Pt: 315.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00237027
CAS Number: 152548-66-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty