Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fmoc-N-Me-Val-OH 95%

Supplier: AMBEED, INC
Warning

A278601-1G A278601-5G A278601-100G A278601-10G A278601-250MG A278601-1G A278601-5G A278601-25G
103813-420EA 6 USD InStock
103813-420 103853-362 76875-258 76875-260 76875-262 77093-288 77093-290 77305-300
Fmoc-N-Me-Val-OH 95%
Fmoc-N-Me-Val-OH
Precaución: for Research Use Only


Formula: C₂₁H₂₃NO₄
Melting Pt: 187…190 °C
Flash Pt: 272.9 °C
Storage Temperature: Ambient
MDL Number: MFCD00153395
CAS Number: 84000-11-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty