Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fmoc-norArg(Pbf)-OH 97%

Supplier: AMBEED, INC
Warning

A218976-1G A218976-5G A218976-100MG A218976-250MG
103818-760EA 325 USD InStock
103818-760 103849-634 77237-608 77237-610
Fmoc-norArg(Pbf)-OH 97%
Fmoc-norArg(Pbf)-OH
Precaución: for Research Use Only


Formula: C₃₃H₃₈N₄O₇S
MW: 634.74 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD15142017
CAS Number: 1313054-32-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty