Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

O-tert-Butyl-N-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-serine 98%

Supplier: AMBEED, INC
Warning

A252216-5G A252216-5G A252216-100G A252216-10G A252216-1G A252216-500G A252216-25G
103851-878EA 6 USD InStock
103851-878 77091-288 77276-342 77276-344 77276-346 77276-348 77304-576
O-tert-Butyl-N-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-serine 98%
O-tert-Butyl-N-[(9H-fluoren-9-ylmethoxy)carbonyl]-L-serine
Precaución: for Research Use Only


Formula: C₂₂H₂₅NO₅
MW: 383.44 g/mol
Melting Pt: 127…130 °C
Flash Pt: 303.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00037127
CAS Number: 71989-33-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty