Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-(9-Fluorenylmethoxycarbonyl)-O-tert-butyl-L-threonine 97%

Supplier: AMBEED, INC
Warning

A415744-5G A415744-100G A415744-10G A415744-25G
103838-344EA 7 USD InStock
103838-344 76841-932 77278-918 77278-920 77307-174
N-(9-Fluorenylmethoxycarbonyl)-O-tert-butyl-L-threonine 97%
N-(9-Fluorenylmethoxycarbonyl)-O-tert-butyl-L-threonine
Precaución: for Research Use Only


Formula: C₂₃H₂₇NO₅
MW: 397.5 g/mol
Melting Pt: 131…134 °C
Flash Pt: 305.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00077075
CAS Number: 71989-35-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty