Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fmoc-Tle-OH 97%

Supplier: AMBEED, INC
Warning

A183190-1G A183190-25G A183190-5G A183190-100G A183190-500G
103815-616EA 6 USD InStock
103815-616 103846-178 77083-774 77083-776 77083-778 77151-680 77151-682
Fmoc-Tle-OH 97%
Fmoc-Tle-OH
Precaución: for Research Use Only


Formula: C₂₁H₂₃NO₄
MW: 353.418 g/mol
Boiling Pt: 554.1 °C (760 mm Hg)
Melting Pt: 124…127 °C
Flash Pt: 288.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00065649
CAS Number: 132684-60-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty