Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fmoc-L-Tyr(Me)-OH 95%

Supplier: AMBEED, INC
Warning

A108968-1G A108968-250MG A108968-1G A108968-5G A108968-10G
103797-418EA 6 USD InStock
103797-418 103823-206 76869-622 77310-038 77310-040 77549-100
Fmoc-L-Tyr(Me)-OH 95%
Fmoc-L-Tyr(Me)-OH
Precaución: for Research Use Only


Formula: C₂₅H₂₃NO₅
MW: 417.47 g/mol
Boiling Pt: 647.8 °C (760 mm Hg)
Flash Pt: 345.6 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00153368
CAS Number: 77128-72-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty