Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Fmoc-L-cyclopropylalanine 95%

Supplier: AMBEED, INC
Warning

A161415-1G A161415-5G A161415-10G A161415-250MG A161415-1G A161415-25G A161415-5G
103805-486EA 13 USD InStock
103805-486 103843-724 76997-794 76997-796 77300-528 77300-530 77300-532
Fmoc-L-cyclopropylalanine 95%
Fmoc-L-cyclopropylalanine
Precaución: for Research Use Only


Formula: C₂₁H₂₁NO₄
MW: 351.402 g/mol
Boiling Pt: 583.5 °C (760 mm Hg)
Flash Pt: 306.7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00798640
CAS Number: 214750-76-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty